Learn More
Fisherbrand™ SQ-EASY™ PreFilter Tips
Disposable, efficient self-contained filtration system for sample preparation applications.
Description
- For use in stool, serum, diluted whole blood, urine, and throat swab assays
- Components are liquid-tight and interlocking to prevent sample leakage
- Range of filtration materials available, including ultra-high molecular weight polyethylene (UHMWPE), high-density polyethylene (HDPE), and glass fiber
- Reagents can be impregnated directly into the filter materials
- All components are available separately to alllow for fllexibility in tube size and particle retention
Specifications
PreFilter™ system | |
100 Pack | |
For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays | |
Sq-Easy™ Filter Tip |
Hydrophilic filter | |
1μm | |
Glass Fiber, UHMWPE |