Learn More
Fisherbrand™ SQ-EASY™ PreFilter Tips
Disposable, efficient self-contained filtration system for sample preparation applications.
 View more versions of this product
Catalog No. 22-267-387
Add to cart
Description
Description
  • For use in stool, serum, diluted whole blood, urine, and throat swab assays
  • Components are liquid-tight and interlocking to prevent sample leakage
  • Range of filtration materials available, including ultra-high molecular weight polyethylene (UHMWPE), high-density polyethylene (HDPE), and glass fiber
  • Reagents can be impregnated directly into the filter materials
  • All components are available separately to alllow for fllexibility in tube size and particle retention
Specifications
Specifications
For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays
5μm
Hydrophilic filter
Sq-Easy™ Filter Tip
PreFilter™ system
1000 Pack
Glass Fiber, UHMWPE