Learn More
Fisherbrand™ SQ-EASY™ PreFilter Tips
Disposable, efficient self-contained filtration system for sample preparation applications.
Manufacturer:  Fisherbrand™ X79840
 View more versions of this product
Catalog No. 22-267-389
Add to cart
Description
Description
  • For use in stool, serum, diluted whole blood, urine, and throat swab assays
  • Components are liquid-tight and interlocking to prevent sample leakage
  • Range of filtration materials available, including ultra-high molecular weight polyethylene (UHMWPE), high-density polyethylene (HDPE), and glass fiber
  • Reagents can be impregnated directly into the filter materials
  • All components are available separately to alllow for fllexibility in tube size and particle retention
Specifications
Specifications
15μm
PreFilter™ system
Sq-Easy™ Filter Tip
HDPE
Hydrophilic filter
For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays
100 Pack