Learn More
Fisherbrand™ SQ-EASY™ PreFilter Tubes
For use in self-contained sample preparation applications.
 View more versions of this product
Catalog No. 22-267-395
Add to cart
Description
Description
Flexibility and tube size allows for sample use according to methodology and volume.
Specifications
Specifications
0.11 oz., 3.5mL
PreFilter™ system
0.11 oz.
Sq-Easy™ Filtration Tube
For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays
100 Pack
3.5mL