Learn More
Fisherbrand™ Staining Control Slides
Allow staining of control and patient samples on the same slide with the same protocol
This item is not returnable.
View return policy
Description
- Red box on the slide identifies the control tissue
- Frosted area provides a positive marking spot
- Superfrost™ Plus treatment secures tissue to the slide throughout the staining process
- Cat. No. 22-042-910 has an extended frosted area, allowing room for the addition of patient information and/or a bar code label
Specifications
Staining Control Slide | |
Standard Frosted Area | |
For automatic stainers |