Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Stainless-Steel Thermometer Probe for Traceable™ Radio-Signal Remote Hygrometer/Thermometer

For testing liquids, air/gases and semisolids

Catalog No. 15-077-52


Add to cart

Description

Description

This Fisherbrand™ Stainless-Steel Thermometer Probe is designed for use with the Traceable™ Radio-Signal Remote Hygrometer/Thermometer. For testing liquids, air/gases and semisolids.

  • Use with 15-077-51
  • Measures 0.12 dia. x 8.5 in.L (0.3 x 22cm)
  • Cable length: 5 ft. (1.5m)

Order Info

Measures 0.12 dia. x 8.5 in. L (0.3 x 22cm). Cable length, 5 ft. (1.5m). Use with 14-648-54 or 15-077-51 sensor modules (above).

Specifications

Specifications

Traceable Radio-Signal Remote Hygrometer/Thermometer
0.3 cm
5 ft.
150 cm
Stainless Steel Probe
0.12 in.
For testing liquids, air/gases and semisolids
22 cm
Stainless Steel
8.5 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit