Fisherbrand™ Sterile Plastic Culture Tubes: Clear Polystyrene

Ideal for tissue culture procedures

 View more versions of this product

Catalog No. 14-956-5A

  Check Availability
Add to Cart

    Specifications

    Color Clear
    For Use With (Application) Ideal for tissue culture procedures
    Includes Screw cap
    Closure Type Screw
    Closure Yes
    Diameter (Metric) Outer 16mm
    Length (Metric) 125mm
    Material Polystyrene
    Product Type Culture Test Tube
    Sterile Yes

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit