Learn More
Fisherbrand™ Sterile Sampling Bags with Regular Tabs
Ends of bag closure are unprotected
Manufacturer:  Fisherbrand™ BPR3070FS
 View more versions of this product
Catalog No. 01-815-21
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Made from heavy-gauge, virgin polyethylene
  • Bags can accommodate solid, semisolid and liquid materials
  • To use, simply tear off top and open bag using pull tabs
  • Insert sample and seal by twirling the bag away from you four times and bending wire ends inward
  • With two round-wire closures
Order Info
Tape does not extend past ends of internal support wires on bag ends.
Specifications
Specifications
7 x 3 in. (17.8 x 7.6cm)
2.5 mil
4 oz.
PE (Polyethylene)
7 in.
3 in.
17.8cm
7.6cm