Learn More
Fisherbrand™ Sterile Sampling Bags with Regular Tabs
Ends of bag closure are unprotected
Specifications
Material Polyethylene
Thickness (English) 2.50 mil
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Price Quantity & Availability  
01-815-20
View Documents
Fisherbrand™
BPR3050FS
5 x 3 in. (12.7 x 7.6cm) 2 oz. Restricted Item View substitute products »
01-815-21
View Documents
Fisherbrand™
BPR3070FS
7 x 3 in. (17.8 x 7.6cm) 4 oz. Restricted Item View substitute products »
Description
Description
  • Made from heavy-gauge, virgin polyethylene
  • Bags can accommodate solid, semisolid and liquid materials
  • To use, simply tear off top and open bag using pull tabs
  • Insert sample and seal by twirling the bag away from you four times and bending wire ends inward
  • With two round-wire closures
Order Info
Tape does not extend past ends of internal support wires on bag ends.
Specifications
Specifications
Polyethylene
2.50 mil