Learn More
Fisherbrand™ Storage File Systems Available on GSA/VA Contract for Federal Government customers only.
The best balance between economy and superior long-term protection
 View more versions of this product
Catalog No. 22-272957
Add to cart
Description
Description
  • Polymer-coated surface resists moisture penetration and ensures long-term storage
  • Each box holds up to 2000 microscope slides or 500 tissue blocks
  • Minimize inventory space — boxes are shipped flat
  • Pressure tested to 300 lb. (136kg) allowing for safe stacking to maximize storage space
  • Unique locking tabs keep stacked boxes safely secured and stable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Storage Filing System
One large box and eight small drawers
30 lb.
500 tissue block
Corrugated cardboard with polymer-coated surface
13.6kg
Documents
Accessories