Fisherbrand™ SURE-TEK™ 2 Biopsy Cassettes Dispenser Packs

Comes with attached lids

 View more versions of this product

Catalog No. 22-048-156

  Check Availability
Add to Cart

    Specifications

    Product Type Biopsy Cassette
    Color Peach
    Includes Attached lid
    For Use With (Equipment) Sure-tek™ 2 cassette

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit