Fisherbrand™ SURE-TEK™ Biopsy Processing/Embedding Cassettes (with Lids)
Specially designed to hold biopsy specimens during the embedding process, as well as in a storage cabinet
 View all options for this product
Catalog No. 15-200-402D
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Writing Face Angle 45°
    Closure Type Separate Lid
    Color Blue
    Material Acetal Polymer
    Description Biopsy Cassette
    • Anterior writing area is at a 45° angle to make the cassette more suitable to be used with certain types of automatic cassette labeling instruments
    • Made of acetal polymer that is totally resistant to the chemical action of histological solvents
    • One-millimeter openings maximize fluid exchange and ensure proper drainage
    • Includes an easy-to-remove disposable lid
    • Available in 11 non-cytotoxic, non-metallic colors
  • Description & Specifications
Related Products
Product Content Feedback