Fisherbrand™ SureFit™ Ergo-F Pipet Tips
Designed specifically for use with Fisherbrand pipetters
 View more versions of this product
Catalog No. 02-681-471
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Volume (Metric) 100 to 1000μL
    Sterility Nonsterile
    Tip Style Nonfilter
    Packaging Reload, 10 x 100
    For Use With (Equipment) Fisherbrand Finnpipettes and Thermo Scientific Finnpipettes
    • Available in three capacities, with or without aerosol barrier
    • Unique wall design provides an extra-strong seal specific to Fisherbrand pipetters
    • Require minimal ejection force
    • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Description & Specifications
Related Products
Product Content Feedback