Learn More
Fisherbrand™ SureFit™ Ergo-G Pipet Tips
Designed specifically for use with Gilson™ Pipetters
 View more versions of this product
Catalog No. 02-681-054
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Ergonomic fit requires less force for tip insertion and ejection
  • Secure, consistent seal
  • Unique wall design provides an extra strong seal specific to Gilson Pipetters
  • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
Gilson™ Pipetman™
10 to 200μL
Sterile
Hinged Rack
Filtered (w/Aerosol barrier at 100μL)
Gilson™ Pipetman™