Fisherbrand™ SureFit™ Ergo-G Pipet Tips
Designed specifically for use with Gilson™ Pipetters
 View more versions of this product
Catalog No. 02-681-051
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Volume (Metric) 10 to 200μL
    Sterility Sterile
    Tip Style Nonfilter
    Packaging Hinged Rack, 10 x 96
    For Use With (Equipment) Gilson Pipetters
    • Ergonomic fit requires less force for tip insertion and ejection
    • Secure, consistent seal
    • Unique wall design provides an extra strong seal specific to Gilson Pipetters
    • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Description & Specifications
Related Products
Product Content Feedback