Learn More
Fisherbrand™ SureGrip™ Pipet Tip Reload System: Tips
Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks
 View more versions of this product
Catalog No. 05-408-190
  Check Availability
Add to cart
Specifications
Description
Easy-to-use: just load the carrier into an empty pipet tip rack (reuse your old racks or purchase separately). Carrier locks onto rack with a quick, press-and-snap action. When empty, recycle or dispose of the carrier and reuse the rack.
  • Compatible with most manufacturers' racks
  • Reduces waste by 80% Certified DNase, RNase, Human DNA and PCR inhibitor free
Pipet Tips in Racks
  • All tips are available racked in sturdy, reusable CellRacks
  • Withstand numerous autoclave cycles
  • Keep tips aligned for pipetter mounting
Several Tip Sizes Available
  • 10µL Pipet tip has graduation marks at 2µL and 10µL
  • 10µL Long Reach tip features extended length (45mm) to reach the bottom of tubes without contaminating pipetter shaft, and graduation marks at 2µL, 5µL and 10µL
  • 200µL Pipet tip features a soft-fit collar for easy mounting and ejection, and graduation marks at 10µL, 50µL and 100µL
  • 1250µL Pipet tip features an extended length (103mm), racked on 9mm centers (multichannel compatible), and graduation marks at 200µL, 500µL and 1000µL
Description & Specifications
Specifications
SureGrip™ Tips
Nonsterile
Yes
Tips in CellRacks
Compatible with most manufacturers' racks
200μL
Natural
Yes
Certified DNase, RNase, human DNA and PCR inhibitor free
Accessories