Fisherbrand™ SureGrip™ Pipet Tip Reload System: Tips

Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks

 View more versions of this product

Catalog No. 05-408-189

  Check Availability
Add to Cart

    Specifications

    Product Type SureGrip™ Tips
    For Use With (Equipment) Compatible with most manufacturers' racks
    Sterility Nonsterile
    Graduated Yes
    Tip Style True tip; soft-fit collar for easy mounting and ejection
    Packaging Tips in Reloads
    Certifications/Compliance Certified DNase, RNase, human DNA and PCR inhibitor free
    Volume (Metric) 200μL
    Color Natural

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit