Fisherbrand™ SureOne Beveled Pipette Tips, Universal Fit: 1-200µL Available on GSA/VA Contract for Federal Government customers only.
Offers a full line of universal fit beveled pipet tips
 View more versions of this product
Catalog No. 02-707-422
  Check Availability
Add to Cart
    Specifications
    Product Type SureOne™ Tip
    Length (English) 2 in.
    Length (Metric) 5cm
    Sterility Sterile
    Tip Style Beveled Ends
    Packaging Hinged Rack
    Certifications/Compliance RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, DNA-free
    Volume (Metric) 1 to 200μL
    Color Yellow
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback