Fisherbrand™ SureOne Ultra Micropoint Pipette Tip, Universal Fit Available on GSA/VA Contract for Federal Government customers only.

Offers a full line of universal fit ultra micropoint pipet tips

 View more versions of this product

Catalog No. 02-681-440

  Check Availability
Add to Cart

    Specifications

    Product Type SureOne™ Ultra Tip
    Packaging Hinged Rack, 10 x 96
    Length (English) 1.25 in.
    Length (Metric) 3.1cm
    For Use With (Equipment) Ultra micropoint pipet
    Sterility Sterile
    Tip Style Micro Point
    Certifications/Compliance RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, DNA-free
    Volume (Metric) 0.1 to 10μL
    Color Natural

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit