Learn More
Fisherbrand™ Syringe Filters - Nonsterile, Durapore™ PVDF Membrane Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
 View more versions of this product
Catalog No. 09-719-001
Add to cart
Description
Description
Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.
  • Hydrophilic membrane binds far less protein than nitrocellulose, nylon or PTFE
  • Ideal for filtration of protein-containing aqueous, mild organic solutions and other pharmaceutical/biological/biotech solutions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
33mm
0.22μm
Syringe Filter
Durapore Hydrophilic Polyvinylidene Fluoride (PVDF)
150/pk.