Fisherbrand™ Syringe Filters: Nylon Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
 View more versions of this product
Catalog No. 09-720-5
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Sterile No
    No. per Pack 100
    Hold up Volume (After Air Purge) Low
    Pore Size 0.22μm
    Type Syringe Filter
    Quantity 100/Pk.
    Diameter (Metric) 13mm
    Material Nylon (NYL)
    Applications Ideal for aqueous/organic/HPLC applications
  • Description & Specifications
Related Products
Product Content Feedback