Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Syringe Filters: Nylon Membrane

Low hold-up volume helps you to recover virtually all of your sample after filtering

 View more versions of this product

Catalog No. 09-720-6

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.

Nylon Membrane

  • Excellent chemical resistance and low protein binding
  • Ideal for aqueous/organic/HPLC applications
  • Hydrophilic

Specifications

Specifications

Non-sterile
13mm
Ideal for aqueous/organic/HPLC applications
0.45μm
100/Pk.
Syringe Filter
Nylon (NYL)
Low
100
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit