Fisherbrand™ Syringe Filters: PTFE Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
 View more versions of this product
Catalog No. 09-720-7
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Hold up Volume (After Air Purge) Low
    Pore Size 0.22μm
    Type Syringe Filter
    Quantity Pack of 100
    Diameter (Metric) 13mm
    Material Polytetrafluoroethylene (PTFE)
    Applications Ideal for degassing/clarifying aqueous and solvent samples
  • Description & Specifications
Related Products
Product Content Feedback