Fisherbrand™ Test Tube Racks with Biohazard Warning Available on GSA/VA Contract for Federal Government customers only.
Easily identified in the lab and sterilization area by bright orange epoxy coating
 View more versions of this product
Catalog No. 14-793-34C
  Check Availability
Add to Cart
    Specifications
    Positions 72.0,72.0
    Product Type Rack, Tube
    Diameter (Metric) Cavity 16mm
    Autoclavable No
    Type Rack, Tube
    Holds 16mm Tubes
    Material Epoxy Coated
    Tube Size 16mm
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback