Learn More
Fisherbrand™ PTFE Starburst Stirring Heads Individual Heads Available on GSA/VA Contract for Federal Government customers only.
Provide better mixing at bottom of vessel with less splashing.
 View more versions of this product
Catalog No. 14-513-78
Add to cart
Description
Description
Provide better mixing at bottom of vessel with less splashing.
  • Shape creates a deep vortex
  • Provides quiet, stable operation
  • Alnico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
PTFE
1.25 in.
32mm
Alnico Magnet
ISO 9001:2008
White
Stir bar
32mm