Fisherbrand™ Thermometers with Fractional-Degree °C Scale

Choose total or partial immersion

 View more versions of this product

Catalog No. 15-000B

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Alert

These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.


    Specifications

    Immersion Length 76mm
    Length (English) Overall 23.8 in.
    Length (Metric) Overall 610mm
    Subdivision 0.1°C,0.1°C
    Temperature Range (Metric) -1° to +101°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit