Learn More
Fisherbrand™ Thin-Wall 0.2mL Microcentrifuge Tube Strips with Attached Caps
Convenient polypropylene PCR reaction strips incorporate individually attached caps
Catalog No. 05-407-8A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Strips of eight integral 0.2mL tubes feature ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer
  • Strips are easily handled, will precisely fit standard well spacing, and can be used with 8-channel handheld pipetters
  • Certified DNA-, RNase-, and DNase-free
  • Available with either flat (needle pierceable) or dome-topped individually attached hinged caps
  • Positive sealing fully protects the contents from evaporation during the entire thermal cycle