Learn More
Thermo Scientific™ Tissue Path™ IV Tissue Cassettes
Contain no jagged edges or uneven surfaces
 View more versions of this product
Catalog No. 22-292442
Add to cart
Description
Description
  • Unique, two-piece molded hinge ensures specimen safety and facilitates easy removal of the cassette cover from the base during the embedding procedure
  • Cover and base can be disassembled and reassembled with ease
  • Cassettes fit all sizes of popular base molds, metal and plastic
  • Three-sided writing surface assures positive identification
  • Slotted design ensures thorough and rapid exchange of processing fluids
  • Resistant to solvents, decalcifying solutions, and compatible with microwave techniques
Order Info
  • Supplied in cases of four handy dispenser boxes, 250 per box
Specifications
Specifications
Slots
Aqua
Plastic
Tissue Cassette