Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Top-Grain Leather Palm Gloves

Top-grain leather provides the longest wear

 View more versions of this product

Catalog No. 19-048-666C



Add to cart

Description

Description

  • Gunn cut with leather covering palm, thumb, index finger and fingertips
  • Offer more resistance to absorption of oils than other split leathers
  • Available with gauntlet cuff or 2.5 in. (6cm) safety cuff

Specifications

Specifications

31.25 x 13.3 cm (12.3 x 5.2 in.)
Brown
Cotton
5.2 in.
0.9 to 1 mm
12.3 in.
Gauntlet
Cotton, Cowhide Leather
Large
13.3 cm
Black, Blue, Red
31.25 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit