Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ 24-Hour Stopwatches

Digital, handheld

 View more versions of this product

Catalog No. 14-649-18


  Check Availability
Add to cart

Specifications

Description

Digital, handheld, for general-purpose applications. Fisherbrand™ Traceable 24-Hour Stopwatches provide single-action timing, time-out, continuous timing, and cumulative split functions.

  • Resolution: 1 second
  • Clock displays time of day to the second, AM/PM, date, and day of the week
  • Rugged ABS plastic case

Certifications

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST™). Calibrated by an ISO 17025 calibration laboratory accredited by A2LA™.

Description & Specifications

Specifications

0.01%
2.37 in.
LCD
0.37 in.
2 Year Silver Oxide battery, shockproof ABS Plastic case and lanyard
1.57cm
0.01 sec.
0.028kg
24 hr.
ISO 17025, A2LA, NIST
6cm
Ideal for lab or plant use
0.93cm
0.62 in.
ABS Plastic
1 oz.
Stopwatch
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit