Learn More
Fisherbrand™ Three-hole Twistit™ Rubber Stoppers
SDP
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
 View more versions of this product
Catalog No. 14-132D
Add to Cart
Description
Description
  • Simple twist-off seals provide convenient one, two, or three-hole use
  • Solid stoppers are vacuum-tested at 30 in. of mercury
  • High resistance to deformation—even after repeated autoclaving
  • Each stopper has its size number in raised figures on top
Specifications
Specifications
3.2 cm
Three Holes Twistit Stopper
3
Natural Rubber
Multi-purpose applications
2.6 cm
6
6
White
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories