Learn More
Fisherbrand™ Two-Way Slant Racks
For incubating liquid cultures on slant during tissue culture processes.
Specifications
Material Epoxy-coated Heavy-gauge Steel Wire
Positions 40
Type Tube Rack
View More Specs
Products 1
Catalog Number Holds Price Quantity & Availability  
Catalog Number Holds Price Quantity & Availability  
14-796-1
View Documents
Tubes (13 to 16 mm) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
For incubating liquid cultures on slant during tissue culture processes.
  • Prepare either 5° or 20° agar slants in same rack
  • Autoclave media in rack
  • While still hot, set rack on one side for 5° tilt, or other for 20° tilt
  • 5° angle usually used for aerobic cultures, 20° for anaerobic
  • 20° tilt also prevents media from drying out during long-term storage
  • White, epoxy-coated, heavy-gauge steel
Specifications
Specifications
Epoxy-coated Heavy-gauge Steel Wire
40
Tube Rack