Fisherbrand™ Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion
 View more versions of this product
Catalog No. 14-171-225
  Check Availability
Add to Cart
    Specifications
    Length (English) 50 ft.
    Color Clear
    Length (Metric) 15m
    Thickness 1/8 in.
    Diameter (English) Outer 9/16 in.
    Diameter (English) Inner 5/16 in.
  • Description & Specifications
Related Products
Product Content Feedback