Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Ultra Flex Clamping Systems

Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.

 View more versions of this product

Catalog No. 02-217-019


  Check Availability
Add to cart

Description

Description

  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers

Specifications

Specifications

For attaching to lab frames or support stands up to 0.75 in.
6.35cm
0.75 in.
Ultra flex clamping system components
2.5 in.
Lab Frame Connector
1.9cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit