Learn More
Fisherbrand™ Ultra Flex Clamping Systems
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
 View more versions of this product
Catalog No. 02-217-008
  Check Availability
Add to cart
Description
Description
  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
Specifications
Thermometer/thermocouple clamp
30.48cm
2.95, 2.72, 0.51 in.
Ultra flex clamping system components
12 in.
Ultra Flex Clamp System
7.49, 6.9, 1.29cm