Learn More
Fisherbrand™ Ultra Stack and Hang Bins
Ideal for use when organizing inventory
 View more versions of this product
Catalog No. 15-000-223
Add to Cart
Description
Description
  • Made using injection molded polypropylene
  • Designed with front, back and side grips for easy handling
  • Built-in rear hanger allows bins to hang from louvered panels 
  • Large moulded front label slots for ID labels for identification of contents
  • Individual dividers maximize flexibility
  • Wide stacking ledges and anti-slide locks prevents forward shifting
  • Resistant to rust and corrosion
  • Autoclavable up to 250°F
  • Withstands extreme cold temperatures
  • Available in different dimensions and various colors
Warranty and Services
One year
Specifications
Specifications
Storage Container
10.87 x 16.5 x 5 in. (27.6 x 41.9 x 12.7 cm)
12.7 cm
27.6 cm
41.9 cm
Clear
5 in.
10.87 in.
16.5 in.