Learn More
Fisherbrand™ Universal Sorbents Get IT Item
Universal, nonselective sorbents can fully saturate in as little as 20 seconds
 View more versions of this product
Catalog No. 19-034-155
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Do not use particulate, socks, or pillows on hydrofluoric acid. Do not use pads, rolls, rugs, or mats on concentrated sulfuric acid and strong caustics, as fabric casings may degrade if contact is lengthy. Refer to Material Safety Data Sheet for details.
Specifications Description & Specifications
Specifications
27 gal.
40.6cm
Universal Sorbent Pad
Polypropylene
45.72cm
16 ft. x 18 in. (40.6 x 45.72cm)
102.2L
16 ft.
Suited for leaks, spills or drips and for routine maintenance, can be used on mild acids and caustics
18 in.
27 gal., 102.2L