Fisherbrand™ Universal Spill Response Drum Kits
All the supplies necessary to safely clean up oils, acids, caustics, solvents, and toxic spills
 View all options for this product
Catalog No. 19-034-969
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Capacity (Metric) Sorbing 162.7 to 276.3L
    Capacity (English) Sorbing 43 to 73 gal.
    Capacity (English) 95 gal.
    Capacity (Metric) 359.5L
    Product Type Drum Kit
    Includes 11 socks, 20 pillows, 50 absorbent pads, 10 disposal bags with ties 25 lb. bag of sorbent
    Capacity 95 gal., 359.5L
    High-density polyethylene drums hold a supply of Fisherbrand™ sorbent socks, pillows, and sorbent powder—and provide safe containment for used sorbents after spill cleanup. Pull-tab opening makes access quick and easy. Drums are highly resistant to impact and environmental stress cracks, and have safety locks and PVC O-rings to assure tight seal.
    • 6 gal. (22.7L) Drum contains two universal pads, five 3 in. x 4 ft. socks, two cartons of sorbent powder, a yellow bag and twist tie, for a total sorbing capacity of approximately 7 gal. (26.5L)
    • 20 gal. (75.7L) Drum contains 50 universal pads, two cartons of sorbent powder, four 3 in. x 4 ft. socks, two pillows, a pair of gloves and goggles, one bag and twist tie, for a total sorbing capacity of approximately 18 gal. (68.1L)
  • Description & Specifications
Related Products
Product Content Feedback