Fisherbrand™ Universal Spill Response Drum Kits Get IT Item
All the supplies necessary to safely clean up oils, acids, caustics, solvents, and toxic spills
 View more versions of this product
Catalog No. 17-206A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Alert
Do not use particulate, socks, or pillows on hydrofluoric acid. Do not use pads or rolls on concentrated sulfuric acid and strong caustics, as fabric casings may degrade if contact is lengthy. Refer to Material Safety Data Sheet for details.
    Specifications
    Capacity (English) Sorbing 10 to 18 gal.
    Capacity (Metric) Sorbing 37.8 to 68.1L
    Capacity 20 gal., 75.7L
    Certifications/Compliance DOT
    Includes 50 Uuniversal pads, two cartons of sorbent powder, four 3 in. x 4 ft. socks, two pillows, a pair of gloves and goggles, one bag and twist tie for a total sorbing capacity of 18 gal.
    Capacity (English) 20 gal.
    Capacity (Metric) 75.7L
    Product Type Drum Kit
  • Description & Specifications
Related Products
Product Content Feedback