Fisherbrand™ Unlined Plastic Caps

Unlined caps

 View more versions of this product

Catalog No. 02-911-856

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Unlined Cap
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Liner Material Unlined
    Packaging Type Packed in 12 bags of 12
    Quantity 144/Cs.
    Shape Unlined Cap
    Color White
    Material Polypropylene
    Certifications/Compliance ISO 9001
    For Use With Bottle
    Neck Finish 89-400
    No. per Case 144 Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit