Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ UriSystem™ DeciSlide

Analyze up to 10 samples simultaneously

$138.72

Specifications

Material Acrylic
Closure Type Integral coverslip
Bottom Shape Flat
Color Optically Clear
For Use With (Application) Urine sedimentation analysis
View More Specs

Products 2
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
14-377-259 Edge
View Documents
Non-Gridded Pack of 100 for $138.72
Case of 10 PK for $1,212.45
Edge
Add to cart
 
23-640-326
View Documents
Gridded This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Made of optically clear acrylic
  • Features an integral cover slip 10 individual chambers
  • Each chamber is sealed on three sides to prevent cross contamination
  • Reduces cross-contamination while standardizing volume per area
Specifications

Specifications

Acrylic
Flat
Urine sedimentation analysis
Integral coverslip
Optically Clear
10 samples
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit