Learn More
Fisherbrand™ Water Bath Clamps
Holds glass tubes and thermometers onto walls of glass water bath.
Description
- Nickel-plated construction
- Built-in holder grip walls of varying thicknesses, up to 9mm (0.3 in.)
- Knurled thumbscrew tightens jaws to hold objects firmly
- Available in small and large size
- Large water bath clamp comes with non-slip vinyl sleeves and fiberglass covers which are used for applications above 100°C
Specifications
Glass tubes and thermometers | |
Small | |
3.5 in. | |
89 mm |