Learn More
Fisherbrand™ Welder's Gloves
Available in shoulder and side styles
 View more versions of this product
Catalog No. 19-059672
Add to cart
Description
Description
  • Cowhide leather welder's gloves in standard and premium grades
  • Fully sock lined for protection and comfort
  • Feature one-piece leather back and welted seams in vulnerable areas
  • Color coded for easy sorting
  • Premium grade is sewn with heat resistant Kevlar™
  • Men's size
Specifications
Specifications
34.25 x 14 cm (13.5 x 5.5 in.)
Hot and heavy material handling
Cotton Flock
Premium grade; Side style
14 cm
Green
34.25 cm
Gauntlet
Cowhide Leather
Large
5.5 in.
1.2 mm
13.5 in.