Learn More
Fisherbrand™ White Polypropylene Caps with PTFE Liner
For use with analytical lab samples, high-purity chemicals, strong acids and solvents. Fisherbrand™ White Polypropylene Caps with PTFE Liners are an economical alternative to Thermoset and Phenolic PTFE lined caps.
 View more versions of this product
Catalog No. 02-912-064
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ISO 9001
White
100/Pk.
13-425
PTFE/Polyethylene
Polypropylene
Bottle