Fisherbrand™ White Polypropylene Caps with PTFE Liners

Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

 View more versions of this product

Catalog No. 02-912-066

  Check Availability
Add to Cart

    Specifications

    Closure Material Plastic
    Closure Size 18/400
    Closure Type Screw cap
    Description Screw cap
    Liner Material PTFE faced PE
    Quantity 500/PK.
    Sterile Yes
    Material Plastic
    Certifications/Compliance ISO 9001
    Color White
    Neck Finish 18-400
    Closure Liner PTFE/Polyethylene
    For Use With Bottle
    No. per Case 100/Pk.

    • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
    • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit