Learn More
Fisherbrand™ White Polypropylene PE Foam/PTFE Cap
Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene PE Foam/PTFE Cap have a PTFE facing, backed by F217 liners.
 View more versions of this product
Catalog No. 02-912-079
  Check Availability
Add to cart
Description
Description
Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.
  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Specifications
Specifications
ISO 9001
PTFE/Polyethylene
Screw cap
144/Cs.
144EA/Cs.
Cap, PE Foam/PTFE Lined
White
PE Foam/PTFE
Round
Polypropylene
White
Polypropylene
Packed in 12 bags of 12
Sterile
Bottle
83/400
83-400
Screw