Fisherbrand™ White Polypropylene Caps with PTFE Liner Available on GSA/VA Contract for Federal Government customers only.

For use with analytical lab samples, high-purity chemicals, strong acids and solvents. Fisherbrand™ White Polypropylene Caps with PTFE Liners are an economical alternative to Thermoset and Phenolic PTFE lined caps.

 View more versions of this product

Catalog No. 02-912-068

  Check Availability
Add to Cart

    Specifications

    Product Type PE Foam/PTFE Lined Cap
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Liner Material PE Foam/PTFE
    Packaging Type Packed in 5 bags of 100
    Quantity 500EA/Cs.
    Shape Round
    Certifications/Compliance ISO 9001
    Closure Liner PTFE/Polyethylene
    Color White
    For Use With Bottle
    Material Polypropylene
    Neck Finish 22-400
    No. per Case 500/Cs.

    • Ideal for environmental samples, pharmaceuticals and diagnostic reagents
    • Screw cap sizes for narrow- and wide-mouth bottles

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit