Learn More
Fisherbrand™ Premium Full-Frosted Cytology Microscope Slides Available on GSA/VA Contract for Federal Government customers only.
With sandblasted frosting over one entire side
Catalog No. 12-544-5CY
Add to cart
Description
Description
With sandblasted frosting over one entire side
  • Frosting disappears when cover slip is mounted over specimen—doesn't interfere with viewing
  • 0.097 to 1.07mm thick
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
76.2 mm
25.4 mm
1 in.
0.97 to 1.07mm
3 in.