Fisherfinest™ Transport Swabs

For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms

 View more versions of this product

Catalog No. 14-907-18

  Check Availability
Add to Cart

    Specifications

    Product Type Transport Swab
    Closure Color Orange
    For Use With (Application) For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms
    Sterile Yes
    Tip Material Rayon
    Format Swab
    Packaging Peel pouch
    Quantity 50 Pack
    Swab Configuration Short wire
    Media Type Amies Charcoal

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit