Learn More
3M™ Fit Test Solution
Meets the performance criteria for fit testing respirators under the current OSHA Standard for Respirator Protection: 29 CFR 1910.134, Appendix A.
Manufacturer:  3M™ 50051138163568
Catalog No. 19-072-006
Add to cart
Description
Description
  • Sweet taste fit test solution for fit testing
  • Used with the 3M™ Qualitative Fit Test Apparatus FT-10 and 3M™ Training and Fit Testing Case FT-20
  • Can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • For Occupational Use Only
Specifications
Specifications
1.86 oz.
55 mL
Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators., The products can be used to check face to respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.
Fit Test Solution
Sweet
55 mL
CE Approved
3M™ Qualitative Fit Test Apparatus FT-10, 3M™ Training and Fit Testing Case FT-20
6/case